Order Entry
Poland
Orders LinkContactUsLinkComponent
Quinaldic acid 99%

Quinaldic acid 99%

Glass bottle, 50 g
Catalog # ACRO171050500
Irritant
Quinaldic acid 99%
Catalog # ACRO171050500
CAS Number:  93-10-7
Quinaldic acid 99%
Quinaldic acid 99%

Glass bottle, 50 g

CAS Number:  93-10-7
Price
1 580 zł
Log in for availability
1 * 50 g
Product Details & Documents
Synonyms:  Quinoline-2-carboxylic acid, 2-Quinolinecarboxylic acid
  • CAS Number:93-10-7
  • Formula:C₁₀H₇NO₂
  • MDL Number:MFCD00006752
  • Molecular Weight:173.17 g/mol
  • Merck Index:12,08227
  • Melting Pt:159…161 °C
  • Storage Temperature:Ambient
  • EINECS:202-218-3

Documents

Specifications

Specification Test Results

  • Appearance:
    Light brown Crystalline powder or needles
  • Infrared spectrum:
    Conforms
  • Melting point:
    157 to 159 °C
  • Titration with NaOH:
    ≥98.5 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.