Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Copper(I) oxide 97%

Glass bottle, 2,5 kg
Catalog # ACRO197420025
Corrosive
Irritant
Environment
Copper(I) oxide 97%
Catalog # ACRO197420025
CAS Number:  1317-39-1
Copper(I) oxide 97%
Copper(I) oxide 97%

Glass bottle, 2,5 kg

CAS Number:  1317-39-1
Price
1 360 zł
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  Cuprous oxide, Dicopper oxide
  • CAS Number:1317-39-1
  • Formula:Cu₂O
  • MDL Number:MFCD00010974
  • Molecular Weight:143.09 g/mol
  • Merck Index:13,02694
  • Boiling Pt:1800 °C (1013 hPa)
  • Melting Pt:1232 °C
  • Density:5,7 to 6.2 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-270-7
  • UN:3077
  • ADR:9,III

 

Specifications

Specification Test Results

  • Appearance:
    Red-brown Powder
  • Titration Complexometric:
    ≥96.0 %
  • Iron (Fe):
    ≤500 ppm
  • Nickel (Ni):
    ≤100 ppm
  • Silver (Ag):
    ≤50 ppm

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.