Order Entry
Poland
Orders LinkContactUsLinkComponent
Lead(II) nitrate 99+%

Lead(II) nitrate 99+%

Plastic bottle, 5 kg
Catalog # ACRO211560051
Irritant
Health
Environment
Oxidizing
Corrosive
Lead(II) nitrate 99+%
Catalog # ACRO211560051
CAS Number:  10099-74-8
Lead(II) nitrate 99+%
Lead(II) nitrate 99+%

Plastic bottle, 5 kg

CAS Number:  10099-74-8
Price
2 060 zł
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  Lead dinitrate
  • CAS Number:10099-74-8
  • Formula:Pb(NO₃)₂
  • MDL Number:MFCD00011153
  • Molecular Weight:331.21 g/mol
  • Merck Index:13,05432
  • Melting Pt:470 °C (dec.)
  • Density:4.53 g/cm³
  • Storage Temperature:Ambient
  • EINECS:233-245-9
  • UN:1469
  • ADR:5.1,II

Documents

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals and/or chunks
  • Titration Complexometric:
    ≥99.0 %
  • Chloride (Cl):
    ≤10 ppm
  • Iron (Fe):
    ≤10 ppm

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.