Benzaldehyde 98+%
Metal drum, 25 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:100-52-7
- Formula:C₆H₅CHO
- MDL Number:MFCD00003299
- Molecular Weight:106.12 g/mol
- Merck Index:13,01057
- Boiling Pt:179 °C (1013 hPa)
- Melting Pt:–26 °C
- Density:1.043 g/cm³
- Flash Pt:64 °C
- Storage Temperature:Ambient
- EINECS:202-860-4
- UN:1990
- ADR:9,III
Documents
ACRO10522
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5440 to 1.5460 (20°C, 589 nm)
- GC:≥98.0 %
- Specific gravity:(25°C) 1.041 to 1.046
- Acid value:≤5 mg KOH/g
Specifications
- Pack type:Metal drum
- Pk:25 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



