Order Entry
Poland
Orders LinkContactUsLinkComponent
 

4'-Nitroacetophenone 97%

Glass bottle, 1 kg
  ACRO128340010
 
4'-Nitroacetophenone 97%
  ACRO128340010
 :  100-19-6
4'-Nitroacetophenone 97%
4'-Nitroacetophenone 97%

Glass bottle, 1 kg

 :  100-19-6
 
918 zł
   
1 * 1 kg
 
 :  p-Nitroacetophenone, 1-(4-Nitrophenyl)ethanone
  •  :100-19-6
  •  :C₈H₇NO₃
  •  :MFCD00007355
  •  :165.15 g/mol
  •  :202 °C (760 mmHg)
  •  :76…80 °C
  •  :149 °C
  •  :Ambient
  •  :202-827-4

 

 

 

  • Appearance:
    Yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    76 to 80 °C
  • Water:
    ≤0.6 %
  • GC:
    ≥96.0 %
  • Free acid:
    ≤0.2 % (as 4-Nitrobenzoic acid)

 

  • Pack type:
    Glass bottle
  • Pk:
    1 kg

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.