Order Entry
Poland
ContactUsLinkComponent
Configuration ,,CapsuleConnections,,,,Hose for 4 to 9.5 mm I.D. latex tubing and Female LuerslipHousing,,,,,,PolypropyleneMaximum Inlet Pressure,,4.1 bar (25°C)Sterility ,,,,Non-SterileSteri 1 * 10 ST
Catalog # MILXMTGR05010
Supplier:  Merck
Configuration ,,CapsuleConnections,,,,Hose for 4 to 9.5 mm I.D. latex tubing and Female LuerslipHousing,,,,,,PolypropyleneMaximum Inlet Pressure,,4.1 bar (25°C)Sterility ,,,,Non-SterileSteri 1 * 10 ST
Catalog # MILXMTGR05010
Supplier:  Merck
Frequently Bought Together