Order Entry
Poland
ContactUsLinkComponent
MidiKros 20cm MPES 5kD 1,0mm FLL X FLL sterile 1 * 1 items
  SPECD02-E005-10-S
MidiKros 20cm MPES 5kD 1,0mm FLL X FLL sterile 1 * 1 items
  SPECD02-E005-10-S
Frequently Bought Together