Order Entry
Poland
Orders LinkContactUsLinkComponent
WHEATON® Diagnostic Vials

WHEATON® Diagnostic Vials

Tubular vials, 20,00 ml, 30×52 mm, Borosilicate glass, Amber, No, 20 - 400
Catalog # WHEAW216377
Supplier:   DWK Life Sciences
WHEATON® Diagnostic Vials
Catalog # WHEAW216377
Supplier:  DWK Life Sciences
WHEATON® Diagnostic Vials
WHEATON® Diagnostic Vials

Tubular vials, 20,00 ml, 30×52 mm, Borosilicate glass, Amber, No, 20 - 400

Price
6 200 zł
Log in for availability
1 * 480 SZT
Product Details & Documents

These vials are manufactured from borosilicate glass that conforms to USP Type I requirements. The screw thread design eliminates the need for crimping tools.

  • Alternative to serum bottles and vials
  • Ideal for lyophilisation
  • Fits with I-Loc™ closure or screw cap with thin flange stopper
  • Heavy walled glass provides impact resistance
Specifications
  • Cap included:
    No
  • Cap Size:
    20 - 400
  • Ø×H:
    30×52 mm
  • Capacity:
    20,00 ml
  • Colour:
    Amber
  • Description:
    Tubular vials
  • Material:
    Borosilicate glass
  • Seq:
    06