Order Entry
Poland
ContactUsLinkComponent
Themometer ASTM, 37C -2+52:0.2C, white backed, mercury filled, immersion 100mm, 395x6-8mm, suitable for govermment verificatio, durable pigment with manufacturere declaration 1 * 1 items
Catalog # AMAR300570KB
Supplier:  Amarell
Themometer ASTM, 37C -2+52:0.2C, white backed, mercury filled, immersion 100mm, 395x6-8mm, suitable for govermment verificatio, durable pigment with manufacturere declaration 1 * 1 items
Catalog # AMAR300570KB
Supplier:  Amarell
Frequently Bought Together