Silver Nitrate 0.01 mol/l (0.01N) volumetric solution 1 * 1.000 mL
Catalog # APPC182564.1211
Supplier: PanReac AppliChem
Silver Nitrate 0.01 mol/l (0.01N) volumetric solution 1 * 1.000 mL
Catalog # APPC182564.1211
Supplier: PanReac AppliChem
Frequently Bought Together
Sodium chloride 99.0-100.5% USP, Ph. Eur., BP
Supplier: VWR Chemicals
Hydrochloric acid 30%, TECHNICAL
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Desiccator lids, DURAN®
Supplier: DWK Life Sciences
Potassium hexacyanoferrate(III) ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Potassium iodide ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
Sulphuric acid 50%, TECHNICAL
Supplier: VWR Chemicals
Sample bags with closure, SteriBag StandUp
Supplier: BURKLE
D003 TTC/RBS DIPSLIDES (ROSE BENGAL) PACK OF 10 1 * 10 items
Supplier: Lovibond
Test kits, photometric, Sulphide, LCK cuvette tests
Supplier: Hach
FAM benzine, 65-95 °C DIN 51635, Riedel-de Haën™
Supplier: Honeywell Chemicals
Silver nitrate 0.1 M AgNO3 (0.1N) in aqueous solution ≥99% Reag. Ph. Eur. volumetric solution, Fluka™
Supplier: Honeywell Chemicals