Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Sodium hypochlorite solution 5% (w/w), Technical Grade

Plastic bottle, 1.000 mL
Catalog # APPC212297.1211
Corrosive
Environment
Supplier:   PanReac AppliChem
Sodium hypochlorite solution 5% (w/w), Technical Grade
Catalog # APPC212297.1211
Supplier:  PanReac AppliChem
CAS Number:  7681-52-9
Sodium hypochlorite solution 5% (w/w), Technical Grade

Plastic bottle, 1.000 mL

CAS Number:  7681-52-9
Price
125 zł
Log in for availability
1 * 1.000 mL
Product Details & Documents
  • CAS Number:7681-52-9
  • Formula:ClNaO
  • MDL Number:MFCD00011120
  • Molecular Weight:74.44 g/mol
  • Storage Temperature:Ambient
  • UN:1791
  • ADR:8,III
Specifications

Specification Test Results

  • Assay (iodometr.):
    5% ± 0.5% (At the moment of the batch analysis)

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1.000 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.