Needle seat, high pressure, PEEK, 0.12 mm, for Agilent G7167 Multisampler 1 * 1 items
Catalog # HEWLG4267-87012
Dostawca: Agilent
Needle seat, high pressure, PEEK, 0.12 mm, for Agilent G7167 Multisampler 1 * 1 items
Catalog # HEWLG4267-87012
Dostawca: Agilent
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Dostawca: VWR Chemicals
Syringe filters, Acrodisc® One, wwPTFE membrane
Dostawca: Cytiva (Formerly Pall Lab)
Water, HiPerSolv CHROMANORM® for HPLC
Dostawca: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dostawca: VWR Chemicals
VWR®, Nitrile Gloves
Dostawca: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dostawca: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Dostawca: VWR Collection
VWR®, Bottles, Wide Neck, with Screw Cap
Dostawca: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dostawca: VWR Chemicals
FILTER DISCS2.1MM ID0.2M QUICK CHANGE 1 * 5 items
Dostawca: Agilent
Assortment Boxes, PP
Dostawca: hünersdorff GmbH
Capillaries for InfinityLab quick turn fittings
Dostawca: Agilent
Capillaries for LC systems
Dostawca: Agilent
Fittings, InfinityLab
Dostawca: Agilent