BUTTON SET PICUS 1 * 1 items
numer katalogowy BHIT736008
Dostawca: Sartorius
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Dostawca: VWR Chemicals
Schutzfilter 5 ml für Eppendorf Research Plus 1 * 50 ST
Dostawca: EPPENDORF
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dostawca: VWR Chemicals
VWR®, Accessories for Mechanical Pipettes
Dostawca: VWR Collection
VWR®, Digital Timer, Four Channels, with Clock
Dostawca: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dostawca: VWR Chemicals
VWR® Measuring Cylinders, Tall Form, Class A, Borosilicate Glass
Dostawca: VWR Collection
VWR®, Nitrile Gloves
Dostawca: VWR Collection
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Dostawca: HIRSCHMANN
VWR®, Transfer Pipettes, High Performance
Dostawca: VWR Collection
Accessories for accu-jet® pro
Dostawca: Brand
VWR® Serological Pipette, Standard Line
Dostawca: VWR Collection
Detergents, liquid, for automated cleaning of glassware, neodisher® FLA
Dostawca: Dr. Weigert
Vacuum seal PTFE / FKM
Dostawca: Heidolph Instruments GmbH & Co.KG
Accessories for water purification millipore
Dostawca: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dostawca: VWR Chemicals
Methanol, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
Dostawca: Avantor