Order Entry
Poland
Orders LinkContactUsLinkComponent
WHEATON® Diagnostic Vials

WHEATON® Diagnostic Vials

Tubular vials, 5,00 ml, 22×38 mm, Borosilicate glass, Amber, No, 20 - 400
Catalog # WHEAW216375
Supplier:   DWK Life Sciences
WHEATON® Diagnostic Vials
Catalog # WHEAW216375
Supplier:  DWK Life Sciences
WHEATON® Diagnostic Vials
WHEATON® Diagnostic Vials

Tubular vials, 5,00 ml, 22×38 mm, Borosilicate glass, Amber, No, 20 - 400

Price
5 680 zł
Log in for availability
1 * 480 SZT
Product Details & Documents

These vials are manufactured from borosilicate glass that conforms to USP Type I requirements. The screw thread design eliminates the need for crimping tools.

  • Alternative to serum bottles and vials
  • Ideal for lyophilisation
  • Fits with I-Loc™ closure or screw cap with thin flange stopper
  • Heavy walled glass provides impact resistance
Specifications
  • Cap included:
    No
  • Cap Size:
    20 - 400
  • Ø×H:
    22×38 mm
  • Capacity:
    5,00 ml
  • Colour:
    Amber
  • Description:
    Tubular vials
  • Material:
    Borosilicate glass
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.