Order Entry
Poland
Orders LinkContactUsLinkComponent
SPE columns, Supra-Poly® HLB

SPE columns, Supra-Poly® HLB

HLB, 30 µm, 6 ml, 200 mg
Catalog # PERCN9306673
Supplier:   PerkinElmer
SPE columns, Supra-Poly® HLB
Catalog # PERCN9306673
Supplier:  PerkinElmer
SPE columns, Supra-Poly® HLB

HLB, 30 µm, 6 ml, 200 mg

Price
3 210 zł
Log in for availability
1 * 1 SZT
Product Details & Documents
  • Contains macro-porous polymers with ultra-pure, pharmaceutical grade spherical particles
  • Shorter analysis times, greater load capacity and reduced solvent usage
  • Ideal for high throughput assays
Specifications
  • Particle Size:
    30 µm
  • Capacity:
    6 ml
  • Seq:
    13
  • Bed weight:
    200 mg
  • Phase:
    HLB
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.