SPE columns, Supra-Poly® HLB
HLB, 30 µm, 3 ml, 60 mg
PERCN9306658
- Contains macro-porous polymers with ultra-pure, pharmaceutical grade spherical particles
- Shorter analysis times, greater load capacity and reduced solvent usage
- Ideal for high throughput assays
- Particle Size:30 µm
- Capacity:3 ml
- Seq:06
- Bed weight:60 mg
- Phase:HLB



