Kolumienki SPE, Supra-Poly® HLB
Supra-Poly HLB column, HLB, 30 µm, 6 ml, 500 mg
Catalog # PERCN9306674
Supplier: PerkinElmer
Jump to:
Product Details & Documents
- Contains macro-porous polymers with ultra-pure, pharmaceutical grade spherical particles
- Shorter analysis times, greater load capacity and reduced solvent usage
- Ideal for high throughput assays
Specifications
- Wymiary cząstek:30 µm
- Pojemność:6 ml
- Opis:Supra-Poly HLB column
- Seq:15
- Waga sorbentu:500 mg
- Faza:HLB



