SPE columns, Supra-Poly® HLB
HLB, 30 µm, 3 ml, 200 mg
Catalog # PERCN9306660
Supplier: PerkinElmer
Jump to:
Product Details & Documents
- Contains macro-porous polymers with ultra-pure, pharmaceutical grade spherical particles
- Shorter analysis times, greater load capacity and reduced solvent usage
- Ideal for high throughput assays
Specifications
- Particle Size:30 µm
- Capacity:3 ml
- Seq:08
- Bed weight:200 mg
- Phase:HLB



