SPE columns, Supra-Poly® HLB
HLB, 30 µm, 3 ml, 500 mg
numer katalogowy PERCN9306661
Dostawca: PerkinElmer
Przejdź do:
Product Details & Documents
- Contains macro-porous polymers with ultra-pure, pharmaceutical grade spherical particles
- Shorter analysis times, greater load capacity and reduced solvent usage
- Ideal for high throughput assays
Dane techniczne
- Particle Size:30 µm
- Capacity:3 ml
- Seq:09
- Bed weight:500 mg
- Phase:HLB
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



