Kolumienki SPE, Supra-Poly® HLB
Supra-Poly HLB column, HLB, 60 µm, 3 ml, 100 mg
Catalog # PERCN9306679
Supplier: PerkinElmer
Jump to:
Product Details & Documents
- Contains macro-porous polymers with ultra-pure, pharmaceutical grade spherical particles
- Shorter analysis times, greater load capacity and reduced solvent usage
- Ideal for high throughput assays
Specifications
- Wymiary cząstek:60 µm
- Pojemność:3 ml
- Opis:Supra-Poly HLB column
- Seq:29
- Waga sorbentu:100 mg
- Faza:HLB



