RTĘCI (II) JODEK CZERWONY CZDA, ACS 1 * 100 g
Catalog # POCH769060113-100G
Supplier: POCH AVANTOR BRAND
RTĘCI (II) JODEK CZERWONY CZDA, ACS 1 * 100 g
Catalog # POCH769060113-100G
Supplier: POCH AVANTOR BRAND
Frequently Bought Together
Barium chloride dihydrate ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Potassium hydrogen phthalate ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ammonium iron(III) sulphate dodecahydrate 99.0-101.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Potassium dihydrogen phosphate ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Zinc sulphate heptahydrate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sodium chloride regeneration salt, TECHNICAL for water softening
Supplier: VWR Chemicals