Bonded PP caps for sample storage vial kits
Bonded white caps with PTFE/silicone liner, 24-414, PP
Catalog # HEWL5183-4308
Supplier: Agilent
Jump to:
Product Details & Documents
Bonded caps include bonded septa that will prevent liner fallout during shipping and sampling. Caps are manufactured of high quality materials to eliminate sample contamination.
Specifications
- Cap Size:24-414
- Description:Bonded white caps with PTFE/silicone liner
- Material:PP
- Seq:08