Order Entry
Poland
Orders LinkContactUsLinkComponent
Bonded PP caps for sample storage vial kits | Avantor
Bonded PP caps for sample storage vial kits
Bonded white caps with PTFE/silicone liner, 20-400, PP
numer katalogowy HEWL5183-4307
Dostawca:   Agilent
Bonded PP caps for sample storage vial kits
numer katalogowy HEWL5183-4307
Dostawca:  Agilent
Bonded PP caps for sample storage vial kits
Bonded PP caps for sample storage vial kits
Bonded white caps with PTFE/silicone liner, 20-400, PP
Przejdź do:
Price
277 zł
Zaloguj się dla dostępności
1 * 100 SZT
Product Details & Documents
Bonded caps include bonded septa that will prevent liner fallout during shipping and sampling. Caps are manufactured of high quality materials to eliminate sample contamination.
Dane techniczne
  • Cap Size:
    20-400
  • Description:
    Bonded white caps with PTFE/silicone liner
  • Material:
    PP
  • Seq:
    07