Ext light path capillary top seller kit includes extended light path capillaries 5 um id,15 um path length. effective length:4,5 and 72 cm 2 capillaries each alignment interface and cybertool 1 * 1 items
Catalog # HEWL5067-4621
Supplier: Agilent
Frequently Bought Together