Crosslab ph combination electrode, non-refillable, polycarbonate (pc) body, bnc connector, glass bulb measuring tip 1 * 1 items
HEWL8010-0972
: Agilent
Crosslab ph combination electrode, non-refillable, polycarbonate (pc) body, bnc connector, glass bulb measuring tip 1 * 1 items
HEWL8010-0972
: Agilent
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection