Caps and seals for wash sta vials(1) 1 * 1 items
Catalog # HEWLG6500-88027
Dostawca: Agilent
Frequently Bought Together
VWR®, Screw Caps for Short Thread Vials ND9
Dostawca: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dostawca: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Dostawca: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dostawca: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dostawca: VWR Chemicals
Acetone ≥99%, TECHNICAL
Dostawca: VWR Chemicals
VWR®, Short Thread Vials ND9
Dostawca: VWR Collection
VWR®, Crimp Caps for Crimp Neck Headspace Vials ND20
Dostawca: VWR Collection
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Dostawca: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dostawca: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dostawca: VWR Chemicals
Headspace crimp top vials, CrossLab
Dostawca: Agilent