Order Entry
Poland
ContactUsLinkComponent
Preconfigured kit isis discrete sampling. suits the isis-ds used with the agilent 77/8 series icp-ms. includes a set of sample lines and connectors for discrete sampling 1 * 1 items
Catalog # HEWLG4911-68202
Supplier:  Agilent
Preconfigured kit isis discrete sampling. suits the isis-ds used with the agilent 77/8 series icp-ms. includes a set of sample lines and connectors for discrete sampling 1 * 1 items
Catalog # HEWLG4911-68202
Supplier:  Agilent
Frequently Bought Together