Order Entry
Poland
ContactUsLinkComponent
Certified headspace crimp top vial, 1 ml, 23 x 46 mm, clear with graduation marks and write-on spot, flat bottom, 1/pk 1 * 100 items
Catalog # HEWL5190-2285
Supplier:  Agilent
undefined
Certified headspace crimp top vial, 1 ml, 23 x 46 mm, clear with graduation marks and write-on spot, flat bottom, 1/pk 1 * 100 items
Catalog # HEWL5190-2285
Supplier:  Agilent
Frequently Bought Together