Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
AMMONIUM MOLYBDATE 4H2O PURE 1 * 250 g
Catalog # CHMP421390000.0250
Supplier:
CHEMPUR
Expand image
AMMONIUM MOLYBDATE 4H2O PURE 1 * 250 g
Catalog # CHMP421390000.0250
Supplier:
CHEMPUR
Frequently Bought Together
Trifluoromethanesulphonic acid for synthesis
Supplier: Merck
Lead ≥99.999% (metals basis), shot 3 mm (0.1 in)
Supplier: Thermo Fisher Scientific
AMMONIUM METAVANADATE 1 * 25 g
Supplier: Merck
Magnesium sulphate heptahydrate, pure analytical reagent
Supplier: CHEMPUR
POTASU CHLORAN CZDA 1 * 500 g
Supplier: CHEMPUR
Iron(III) chloride hexahydrate ≥99%, Puriss. p.a. Reag. Ph. Eur., Fluka™
Supplier: Honeywell Chemicals