Order Entry
Poland
ContactUsLinkComponent
Silica x 3µm particle size x 120Å pore size, 150mm length x 2.1mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2SIL-03N02P
Silica x 3µm particle size x 120Å pore size, 150mm length x 2.1mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2SIL-03N02P
Frequently Bought Together