Order Entry
Poland
ContactUsLinkComponent
C18Q x 5µm particle size x 100Å pore size, 50mm length x 300µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C183-05L30K
C18Q x 5µm particle size x 100Å pore size, 50mm length x 300µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C183-05L30K
Frequently Bought Together