Order Entry
Poland
ContactUsLinkComponent
C8H x 5µm particle size x 120Å pore size, 100mm length x 150µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C83-05M15K
C8H x 5µm particle size x 120Å pore size, 100mm length x 150µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C83-05M15K
Frequently Bought Together