Order Entry
Poland
ContactUsLinkComponent
TRAY CHIP 4 INCH Pocket size: 6,7 x 8,9 x 0,4mm STAT PRO400 dark green = -74D05D All trays with bumps on backside of the tray H44-264350-74D05D 1 * 50 ST
Catalog # ENTEH4426435074D5D
Supplier:  ENTEGRIS
undefined
TRAY CHIP 4 INCH Pocket size: 6,7 x 8,9 x 0,4mm STAT PRO400 dark green = -74D05D All trays with bumps on backside of the tray H44-264350-74D05D 1 * 50 ST
Catalog # ENTEH4426435074D5D
Supplier:  ENTEGRIS
Frequently Bought Together