Order Entry
Poland
Orders LinkContactUsLinkComponent
PolySat® Pre-Saturated Wipes | Avantor

PolySat® Pre-Saturated Wipes

TX1051, 280×230 mm, 50 wipes/flexpack
numer katalogowy 115-2829
Dostawca:   Texwipe
PolySat® Pre-Saturated Wipes
numer katalogowy 115-2829
Dostawca:  Texwipe
PolySat® Pre-Saturated Wipes
PolySat® Pre-Saturated Wipes

TX1051, 280×230 mm, 50 wipes/flexpack

Przejdź do:
Price
Zapytaj o cenę
Zaloguj się dla dostępności
1 * 50 SZT
Product Details & Documents

PolySat® is constructed from a 100% melt-blown polypropylene substrate that is pre-wetted with 70% isopropyl alcohol and packaged in a resealable FlexPack. PolySat® is convenient for cleaning sensitive surfaces and polishing equipment in controlled environments.

  • FlexPacks are a convenient alternative to wipes and spray bottles, eliminating mixing, sterilising and waste
  • Packed in SolventSafe plastic that can be wiped down prior to introduction into the cleanroom
  • Delivers a pre-measured amount of high purity alcohol to the surface: Optimises alcohol usage and reduces spill potential
  • Low particle generation and low levels of solvent extractables

Colour: White

Uwaga: The product(s) on this page are outside the scope of the EU Directive 2019/904.

Dane techniczne
  • Packed:
    50 wipes/flexpack
  • Description:
    TX1051
  • L×W:
    280×230 mm
  • Seq:
    1