Order Entry
Poland
Orders LinkContactUsLinkComponent
GC septa, Marathon

GC septa, Marathon

Marathon septa, 9 mm
Catalog # 555-8184
GC septa, Marathon
Catalog # 555-8184
GC septa, Marathon
GC septa, Marathon

Marathon septa, 9 mm

Price
773 zł
Log in for availability
1 * 50 SZT
Product Details & Documents

Made of low-bleed silicone, these septa have excellent mechanical properties and are ideal for demanding GC-MS applications.

  • Pre-pierced for reliable performance
  • Op to 400 injections per septa
  • Maximum temperature 400 °C
Specifications
  • Ø:
    9 mm
  • Description:
    Marathon septa
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.