Order Entry
Poland
Orders LinkContactUsLinkComponent
Filter paper for special applications
480, 70 mm
  BINZ146320
 :   Ahlstrom
Filter paper for special applications
  BINZ146320
 :  Ahlstrom
Filter paper for special applications
Filter paper for special applications
480, 70 mm
93,20 zł
1 * 100 SZT
These filter papers for suitable for special applications.
Grade 11: Plain, hardened.
Grade 389F: Plain, for determination of fat in foodstuffs.
Grade 480: Plain, phase separation paper, hydrophobic silicone-impregnated paper.
Grade 918: Black filter paper, stained with a sulfur colouring to reveal the particles of bright colour, detection of fluorine or silicon and mycelium in cultivated mushrooms.
Grade 470: Plain, kieselguhr (diatomite) paper for fine turbidity.
Grade 69/K: Plain, activated carbon, used in laboratory: clarification and brightening of dull and dark urines, for polarimetric sugar determination and industry: filtration of galvanic baths and clarification of colored liquids.
  • Grade:
    480
  • Seq:
    11
  • Size:
    70 mm