Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Nessler reagent, tablets

DPD 1, 250 Tab.
Catalog # LOVI511231BT
Supplier:   Lovibond
Nessler reagent, tablets
Catalog # LOVI511231BT
Supplier:  Lovibond
CAS Number:  7783-33-7
Nessler reagent, tablets
Nessler reagent, tablets

DPD 1, 250 Tab.

CAS Number:  7783-33-7
Price
458 zł
Log in for availability
1 * 250 Tablet
Product Details & Documents
Synonyms:  Potassium mercuric Iodide, Potassium tetraiodomercurate, Nota 1, Mercury (II) potassium iodide, Potassium tetraiodomercurate – Potassium hydroxide solution
  • CAS Number:7783-33-7
  • MDL Number:MFCD00049656

 

Specifications

Specifications

  • Pk:
    250 Tab.
  • Pk Info:
    DPD 1

Specifications