USP System Suitability Solution 1,4-Benzoquinone (0.5mg/L C) acidified with HCl 1 * 40 mL
Catalog # REGERTOCRSSA
Supplier: REAGECON
USP System Suitability Solution 1,4-Benzoquinone (0.5mg/L C) acidified with HCl 1 * 40 mL
Catalog # REGERTOCRSSA
Supplier: REAGECON
Frequently Bought Together
Buffer solutions in sachets
Supplier: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Buffer solutions as per DIN 19266-01: 2000
Supplier: WTW
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH Standard Solutions, IUPAC, Certified
Supplier: Hach
Buffer solutions in sachets, perpHect® Pouches, Orion™
Supplier: Thermo Orion