Lactobionic acid 97%
Bottle, 250 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Soluble in water
Synonyms: (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanoic acid, 4-O-β-D-Galactopyranosyl-D-gluconic acid
- CAS Number:96-82-2
- Formula:C₁₂H₂₂O₁₂
- MDL Number:MFCD00078147
- Molecular Weight:358.30 g/mol
- Merck Index:14,5342
- Melting Pt:113…118 °C
- Storage Temperature:Ambient
- EINECS:202-538-3
Documents
Specifications
Specification Test Results
- Colour:White to Off-White
- Form:Powder
- Assay (Titration):97% min
- Water Content (Karl Fischer Titration):5,0% max
Specifications
- Pack type:Bottle
- Pk:250 g



