Lactobionic acid 97%
Bottle, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Soluble in water
Synonyms: (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanoic acid, 4-O-β-D-Galactopyranosyl-D-gluconic acid
- CAS Number:96-82-2
- Formula:C₁₂H₂₂O₁₂
- MDL Number:MFCD00078147
- Molecular Weight:358.30 g/mol
- Merck Index:14,5342
- Melting Pt:113…118 °C
- Storage Temperature:Ambient
- EINECS:202-538-3
Documents
Specifications
Specification Test Results
- Colour:White to Off-White
- Form:Powder
- Assay (Titration):97% min
- Water Content (Karl Fischer Titration):5,0% max
Specifications
- Pack type:Bottle
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



