Order Entry
Poland
Orders LinkContactUsLinkComponent
Disposable Post Mortem scalpels
Disposable Post Mortem scalpels
PM40B, with bull nose blade
Catalog # SWAN2581
Supplier:   SWANN MORTON
Disposable Post Mortem scalpels
Catalog # SWAN2581
Supplier:  SWANN MORTON
Disposable Post Mortem scalpels
Disposable Post Mortem scalpels
PM40B, with bull nose blade
Price
524 zł
Log in for availability
1 * 5 SZT
Product Details & Documents
Carbon blade. Non sterile, disposable histopathology and autopsy knives.
  • With easy to remove guard to protect the cutting edge: Users do not have to fix the blade to the handle
  • Available with either standard PM40 blade or PM40B bull nose blade fixed in a distinctive blue handle
Documents
Specifications
  • Type:
    PM40B, with bull nose blade