Order Entry
Poland
ContactUsLinkComponent
Human LCHN full-length ORF ( AAH12493.1, 12 A.A. - 43 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00057189-P012
Dostawca:  Abnova
Human LCHN full-length ORF ( AAH12493.1, 12 A.A. - 43 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00057189-P012
Dostawca:  Abnova
Frequently Bought Together