Human MSRA partial ORF ( NP_036463.1, 1 A.A. - 100 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 µG
ABNOH00004482-Q012
: Abnova
Human MSRA partial ORF ( NP_036463.1, 1 A.A. - 100 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 µG
ABNOH00004482-Q012
: Abnova
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection