RTĘCI (II) CHLOREK CZDA, ACS, ODCZ. FP 1 * 100 g
Catalog # POCH768670117-100G
Supplier: POCH AVANTOR BRAND
RTĘCI (II) CHLOREK CZDA, ACS, ODCZ. FP 1 * 100 g
Catalog # POCH768670117-100G
Supplier: POCH AVANTOR BRAND
Frequently Bought Together
Potassium nitrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium chloride ≥98%, TECHNICAL
Supplier: VWR Chemicals
Potassium hexacyanoferrate(II) trihydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Potassium iodide ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium sulphate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Iron(III) chloride, anhydrous, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium sulphate, anhydrous 98.5-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Sodium hydrogen carbonate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Tin(II) chloride dihydrate ≥98%, AnalaR NORMAPUR® analytical reagent (max. 0.05 ppm Hg)
Supplier: VWR Chemicals
Sodium hexanitritocobaltate(III), Supelco®
Supplier: Merck
Barium chloride dihydrate ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals