Order Entry
Poland
Orders LinkContactUsLinkComponent
Magnesium, turnings

Magnesium, turnings

Plastic bottle for solids, 50 g
Catalog # CHMP526111850.0050
Flammable
Supplier:   CHEMPUR
Magnesium, turnings
Catalog # CHMP526111850.0050
Supplier:  CHEMPUR
CAS Number:  7439-95-4
Magnesium, turnings

Plastic bottle for solids, 50 g

CAS Number:  7439-95-4
Price
123 zł
Log in for availability
1 * 50 g
Product Details & Documents
  • CAS Number:7439-95-4
  • Formula:Mg
  • MDL Number:MFCD00085308
  • Molecular Weight:24.3 g/mol
  • Storage Temperature:Ambient
  • EINECS:231-104-6
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    50 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.