Order Entry
Poland
Orders LinkContactUsLinkComponent
Copper(I) oxide ≥97%

Copper(I) oxide ≥97%

500 g
Catalog # A14436.36
Irritant
Environment
Copper(I) oxide ≥97%
Catalog # A14436.36
CAS Number:  1317-39-1
Copper(I) oxide ≥97%
Copper(I) oxide ≥97%

500 g

CAS Number:  1317-39-1
Price
340 zł
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 97% CAS No.: 1317-39-1 Molecular Formula: Cu2O Formula Weight: 143.08 Odour: Odourless Melting Point: 1235° Boiling Point: 1800° dec. Merck Reference: 14,2664 UN No.: 3077 MDL No.: MFCD00010974

Synonyms:  Cuprous oxide, Dicopper oxide
  • CAS Number:1317-39-1
  • Formula:Cu₂O
  • MDL Number:MFCD00010974
  • Molecular Weight:143.09 g/mol
  • Merck Index:14,2664
  • Boiling Pt:1800 °C (dec.)
  • Melting Pt:1235 °C
  • Density:6.0
  • Storage Temperature:Ambient
  • EINECS:215-270-7
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pk:
    500 g

Specifications