Order Entry
Poland
Orders LinkContactUsLinkComponent
Copper(I) oxide ≥97%

Copper(I) oxide ≥97%

Glass bottle for solids, 2.500 g
Catalog # A14436.0E
Irritant
Environment
Copper(I) oxide ≥97%
Catalog # A14436.0E
CAS Number:  1317-39-1
Copper(I) oxide ≥97%
Copper(I) oxide ≥97%

Glass bottle for solids, 2.500 g

CAS Number:  1317-39-1
Price
1 300 zł
Log in for availability
1 * 2.500 g
Product Details & Documents

Purity: 97% CAS No.: 1317-39-1 Molecular Formula: Cu2O Formula Weight: 143.08 Odour: Odourless Melting Point: 1235° Boiling Point: 1800° dec. Merck Reference: 14,2664 UN No.: 3077 MDL No.: MFCD00010974

Synonyms:  Cuprous oxide, Dicopper oxide
  • CAS Number:1317-39-1
  • Formula:Cu₂O
  • MDL Number:MFCD00010974
  • Molecular Weight:143.09 g/mol
  • Merck Index:14,2664
  • Boiling Pt:1800 °C (dec.)
  • Melting Pt:1235 °C
  • Density:6.0
  • Storage Temperature:Ambient
  • EINECS:215-270-7
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2.500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.