Order Entry
Poland
ContactUsLinkComponent
Spectroscopy cell, semi-micro with ptfe stopper, quartz suprasil, light path: 20mm. spectroscopy cell, semi-micro with ptfe stopper , quartz suprasil, light path: 20mm, outside dim.: 46h x 12.5w x 22.5d mm, inside width: 4.0mm, base thickness: 3.2m 1 * 2 items
Catalog # PERCB0631057
Supplier:  PerkinElmer
Spectroscopy cell, semi-micro with ptfe stopper, quartz suprasil, light path: 20mm. spectroscopy cell, semi-micro with ptfe stopper , quartz suprasil, light path: 20mm, outside dim.: 46h x 12.5w x 22.5d mm, inside width: 4.0mm, base thickness: 3.2m 1 * 2 items
Catalog # PERCB0631057
Supplier:  PerkinElmer
Frequently Bought Together