Order Entry
Poland
Orders LinkContactUsLinkComponent
Trays, SteriWare®

Trays, SteriWare®

Sterile, 3000 ml, 265×310×55 mm
Catalog # SAMP8081P-3000S
Supplier:   Sampling Systems
Trays, SteriWare®
Catalog # SAMP8081P-3000S
Supplier:  Sampling Systems
Trays, SteriWare®
Trays, SteriWare®

Sterile, 3000 ml, 265×310×55 mm

Price
2 050 zł
Log in for availability
1 * 30 SZT
Product Details & Documents

Trays made of PP, translucent.

  • Assembled and packed in a cleanroom
  • Individually bagged
  • FDA & EU 10/2011 conforming materials
  • BSE/TSE free
  • Full batch traceability
  • Available sterile (gamma irradiated)

The disposable tray is ideal for hygienic applications within hospitals and pharmaceutical factories.

Specifications
  • Capacity:
    3000 ml
  • Description:
    Sterile
  • Seq:
    02
  • W×D×H:
    265×310×55 mm