Order Entry
Poland
ContactUsLinkComponent
recombinant/native protein, efnb3 (human) recombinant protein, human efnb3 (np_001397, 28 a.a. - 226 a.a.) partial recombinant prote in with his tag expressed in escherichia coli. 1 * 100 µG
Catalog # ABNOP5856
Supplier:  Abnova
recombinant/native protein, efnb3 (human) recombinant protein, human efnb3 (np_001397, 28 a.a. - 226 a.a.) partial recombinant prote in with his tag expressed in escherichia coli. 1 * 100 µG
Catalog # ABNOP5856
Supplier:  Abnova